missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-butynoate, 99%, Thermo Scientific Chemicals

Product Code. 10165777 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10165777

Brand: Thermo Scientific Acros 430030010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4341-76-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00015182
InChI Key FCJJZKCJURDYNF-UHFFFAOYSA-N
Synonym ethyl 2-butynoate, ethyl tetrolate, 2-butynoic acid ethyl ester, but-2-ynoic acid ethyl ester, 2-butynoic acid, ethyl ester, tetrolic acid ethyl ester, ethyl-2-butynoate, ethyl 3-methylpropiolate, acmc-1aeo2, ethyl 1-propynecarboxylate
PubChem CID 78043
IUPAC Name ethyl but-2-ynoate
SMILES CCOC(=O)C#CC

Specifications

Color Brown to Gray
Boiling Point 160.0°C to 164.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 112.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Ethyl 2-butynoate
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 2-butynoate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.