missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-6-methylpyridine-4-carboxylate, 97%, Thermo Scientific™

Produktkod. 15447725
Change view
Klicka för att se tillgängliga alternativ
Quantity:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15447725 250 mg 250mg
11352959 1 g 1g
11362959 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15447725 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H27778.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 3998-88-7
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.634
MDL Number MFCD09842151
InChI Key UMAAIDIASQFLTF-UHFFFAOYSA-N
Synonym ethyl 2-chloro-6-methylisonicotinate, 2-chloro-6-methylpyridine-4-carboxylic acid ethyl ester, 2-chloro-6-methylisonicotinic acid ethyl ester, ethyl 2-chloro-6-methyl-4-pyridinecarboxylate, ethyl2-chloro-6-methylpyridine-4-carboxylate, 2-chloro-6-methyl-isonicotinic acid ethyl ester, 4-pyridinecarboxylicacid, 2-chloro-6-methyl-, ethyl ester, 2-chloro-6-methylpyridine-4-carboxylic acid ethyl ester; ethyl 2-chloro-6-methylisonicotinate
PubChem CID 22625009
IUPAC Name ethyl 2-chloro-6-methylpyridine-4-carboxylate
SMILES CCOC(=O)C1=CC(=NC(=C1)Cl)C

Specifikationer

Melting Point 65°C to 67°C
Boiling Point 107°C (1.5 mmHg)
Quantity 250 mg
Formula Weight 199.64
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-6-methylpyridine-4-carboxylate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.