missing translation for 'onlineSavingsMsg'
Learn More

ethyl 2-cyanoacetate, Maybridge

Product Code. 10198204
Change view
Click to view available options
:
Amber Glass Bottle; 100g
Product Code.
10198204 Amber Glass Bottle; 100g
1 options
This item is not returnable. View return policy

Product Code. 10198204

Brand: Thermo Scientific Maybridge SB00844FA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 105-56-6
Density 1.0600g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Molecular Formula C5H7NO2
Linear Formula NCCH2CO2C2H5
MDL Number MFCD00001940
Quantity 100 g
Synonym ethyl cyanoacetate, ethyl cyanacetate, cyanoacetic acid ethyl ester, acetic acid, cyano-, ethyl ester, ethyl cyanoethanoate, cyanoacetic ester, cyanacetate ethyle, usaf kf-25, estere cianoacetico, ethyl cyano acetate
InChI Key ZIUSEGSNTOUIPT-UHFFFAOYSA-N
SMILES CCOC(=O)CC#N
IUPAC Name ethyl 2-cyanoacetate
Molecular Weight (g/mol) 113.12
PubChem CID 7764
Formula Weight 113.12
Percent Purity 97%
Chemical Name or Material Ethyl 2-cyanoacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.