missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-ethylacetoacetate, 90%

Product Code. 11925111 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11925111 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11925111 Supplier Thermo Scientific Acros Supplier No. 345840250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 607-97-6 | C8H14O3 | 158.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 607-97-6
Color Colorless to Yellow
Density 0.9810g/mL
Boiling Point 189.0°C (743.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O3
Linear Formula CH3C(O)CH(C2H5)COOC2H5
Quantity 25 g
Specific Gravity 0.981
Synonym ethyl 2-ethylacetoacetate, ethyl 2-acetylbutyrate, butanoic acid, 2-ethyl-3-oxo-, ethyl ester, 2-ethylacetoacetic acid ethyl ester, ethyl 2-ethyl-3-ketobutyrate, ethyl alpha-ethylacetoacetate, ethyl-2-ethylacetoacetate, 2-ethyl-3-oxo-butyric acid ethyl ester, ethyl .alpha.-acetylbutyrate, ethyl 2-ethyl-3-oxo-butanoate
InChI Key OKANYBNORCUPKZ-UHFFFAOYSA-N
SMILES CCC(C(=O)C)C(=O)OCC
IUPAC Name ethyl 2-ethyl-3-oxobutanoate
Molecular Weight (g/mol) 158.2
PubChem CID 79065
Formula Weight 158.2
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethyl 2-ethylacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.