missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-furoate, 99%

Product Code. 11421628
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421628 500 g 500g
11401628 25 g 25g
11411628 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421628 Supplier Thermo Scientific Alfa Aesar Supplier No. A13909.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 614-99-3 | C7H8O3 | 140.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-99-3
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003237
InChI Key NHXSTXWKZVAVOQ-UHFFFAOYSA-N
PubChem CID 11980
IUPAC Name ethyl furan-2-carboxylate
SMILES CCOC(=O)C1=CC=CO1

Specifications

Melting Point 31°C to 36°C
Density 1.12
Boiling Point 195°C to 196°C
Flash Point 70°C (158°F)
Quantity 500 g
Beilstein 2653
Merck Index 14,4307
Synonym ethyl 2-furoate, ethyl furoate, ethyl 2-furancarboxylate, 2-furancarboxylic acid, ethyl ester, ethyl pyromucate, 2-carboethoxyfuran, furan-2-carboxylic acid ethyl ester, ethyl-2-furoate, 2-furoic acid, ethyl ester, ethyl2-furoate
Formula Weight 140.14
Percent Purity 99%
Chemical Name or Material Ethyl 2-furoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.