missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-Mercaptoacetate, TRC

Product Code. 30369841
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30369841 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30369841 Supplier Toronto Research Chemicals Supplier No. TRCE92499510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 2-Mercaptoacetate
CAS 623-51-8
Molecular Formula C4H8O2S
Quantity 10 g
Synonym Mercaptoacetic Acid Ethyl Ester, 2-Mercaptoacetic Acid Ethyl Ester, Ethoxycarbonylmethanethiol, Ethyl Mercaptoacetate, Ethyl Thioglycolate, Ethyl alpha-Mercaptoacetate, Ethylthiomethyl Acetate, Mercaptoacetic Acid Ethyl Ester, NSC 8834, Sulfanylacetic Acid Ethyl Ester, Thioglycolic Acid Ethyl Ester
InChI Formula InChI=1S/C4H8O2S/c1-2-6-4(5)3-7/h7H,2-3H2,1H3
SMILES CCOC(=O)CS
IUPAC Name ethyl 2-sulfanylacetate
Molecular Weight (g/mol) 120.17
Formula Weight 120.0245
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.