missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 11340617
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11340617 250 mg 250mg
10634405 1 g 1g
10462854 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11340617 Supplier Thermo Scientific Maybridge Supplier No. CC42223CB
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 32043-95-1
Molecular Formula C13H13NO2S
Molecular Weight (g/mol) 247.31
MDL Number MFCD07346320
InChI Key UHLMXNFHHFDVPW-UHFFFAOYSA-N
Synonym ethyl 2-methyl-4-phenylthiazole-5-carboxylate, 2-methyl-4-phenylthiazole-5-carboxylic acid ethyl ester, 2-methyl-4-phenyl-thiazole-5-carboxylic acid ethyl ester, 5-thiazolecarboxylicacid, 2-methyl-4-phenyl-, ethyl ester
PubChem CID 7131013
IUPAC Name ethyl 2-methyl-4-phenyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(C)S1)C1=CC=CC=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 247.32
Percent Purity 97%
Chemical Name or Material Ethyl 2-methyl-4-phenyl-1, 3-thiazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.