missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-methylacetoacetate, 95%

Product Code. 11484723
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484723 500 g 500g
11464723 25 g 25g
11474723 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484723 Supplier Thermo Scientific Alfa Aesar Supplier No. A11629.36
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 609-14-3 | C7H12O3 | 144.17 g/mol

Ethyl 2-methylacetoacetate is used in Japp-Klingemann reaction to prepare hydrazones from beta-keto acid and aryl diazonium salts. It is used as a precursor involved in the synthesis of ethylpyruvate phenylhydrazone via reaction with benzenediazonium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-methylacetoacetate is used in Japp-Klingemann reaction to prepare hydrazones from beta-keto acid and aryl diazonium salts. It is used as a precursor involved in the synthesis of ethylpyruvate phenylhydrazone via reaction with benzenediazonium chloride.

Solubility
Miscible with alcohol and ether. Silghtly miscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 609-14-3
Molecular Formula C7H12O3
Molecular Weight (g/mol) 144.17
MDL Number MFCD00009164
InChI Key FNENWZWNOPCZGK-UHFFFAOYSA-N
Synonym ethyl 2-methylacetoacetate, ethyl 2-acetylpropionate, ethyl-2-methyl acetoacetate, ethyl methylacetoacetate, ethyl 2-methyl-3-oxobutyrate, ethyl2-methylacetoacetate, butanoic acid, 2-methyl-3-oxo-, ethyl ester, 2-methylacetoacetic acid ethyl ester, ethyl alpha-methylacetylacetate, 2-methyl-3-oxobutanoic acid ethyl ester
PubChem CID 701
IUPAC Name ethyl 2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C

Specifications

Melting Point -45°C
Density 1.019
Boiling Point 185°C
Flash Point 62°C (143°F)
Odor Ester-like
Refractive Index 1.418
Quantity 500 g
Beilstein 1071742
Solubility Information Miscible with alcohol and ether. Silghtly miscible with water.
Formula Weight 144.17
Percent Purity 95%
Chemical Name or Material Ethyl 2-methylacetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.