missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-morpholino-1,3-thiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10595764
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10595764 5 g Amber Glass Bottle 5g
10092174 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10595764 Supplier Thermo Scientific Maybridge Supplier No. CC62823DE
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 126533-95-7
Molecular Formula C10H14N2O3S
Molecular Weight (g/mol) 242.293
MDL Number MFCD09702412
InChI Key OINWWRRVBHJAKO-UHFFFAOYSA-N
Synonym ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate, ethyl 2-4-morpholinyl thiazole-4-carboxylate, ethyl 2-morpholino-1,3-thiazole-4-carboxylate, ethyl 2-morpholinothiazole-4-carboxylate, 4-thiazolecarboxylicacid, 2-4-morpholinyl-, ethyl ester, acmc-1c0vg, 4-4-ethoxycarbonyl-1,3-thiazol-2-yl morpholine, 4-thiazolecarboxylicacid,2-4-morpholinyl-,ethyl ester, 2-morpholin-4-yl-thiazole-4-carboxylic acid ethyl ester
PubChem CID 15053544
IUPAC Name ethyl 2-morpholin-4-yl-1,3-thiazole-4-carboxylate
SMILES CCOC(=O)C1=CSC(=N1)N2CCOCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 242.3
Percent Purity 97%
Chemical Name or Material Ethyl 2-morpholino-1, 3-thiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.