missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-n-butylacetoacetate, 95%, Thermo Scientific™

Product Code. 15498779
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498779 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15498779 Supplier Thermo Scientific Alfa Aesar Supplier No. L00905.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1540-29-0
Density 0.95
Boiling Point 224°C
Molecular Formula C10H18O3
Refractive Index 1.429
MDL Number MFCD00026888
Quantity 5 g
Beilstein 636718
Synonym ethyl 2-n-butylacetoacetate, ethyl 2-butylacetoacetate, hexanoic acid, 2-acetyl-, ethyl ester, ethyl .alpha.-butylacetoacetate, 2-acetylhexanoic acid ethyl ester, ethyl butyl acetoacetate, ethyl 2-acetyl hexanoate, acmc-209da5, ethyl-2-n-butylacetoacetate, ksc494i1n
InChI Key ZTOQBHVLCJERBS-UHFFFAOYSA-N
SMILES CCCCC(C(=O)C)C(=O)OCC
IUPAC Name ethyl 2-acetylhexanoate
Molecular Weight (g/mol) 186.251
PubChem CID 95466
Formula Weight 186.25
Percent Purity 95%
Chemical Name or Material Ethyl 2-n-butylacetoacetate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.