missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-nitropropionate, 96%

Product Code. 11398705
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398705 1 g 1g
11308715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398705 Supplier Thermo Scientific Alfa Aesar Supplier No. L12261.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2531-80-8 | C5H9NO4 | 147.13 g/mol

Ethyl 2-nitropropionate is used in the preparation of (-)-trans-2,5-dimethylprolineethyl ester via enantioselective Michael reaction methyl vinyl ketone using modified dihydroquinine as catalyst, novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([15N]EMPO) for detecting superoxide anion, 2-ethoxycarbonyl-2-nitropentanal. It is used for enantioselective Michael reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-nitropropionate is used in the preparation of (-)-trans-2,5-dimethylprolineethyl ester via enantioselective Michael reaction methyl vinyl ketone using modified dihydroquinine as catalyst, novel nitrogen-15-labeled nitrone spin trap, 5-ethoxycarbonyl-5-methyl-1-pyrroline N-oxide ([15N]EMPO) for detecting superoxide anion, 2-ethoxycarbonyl-2-nitropentanal. It is used for enantioselective Michael reactions.

Solubility
Not miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2531-80-8
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00024818
InChI Key ZXBGJDZWJJFFQY-UHFFFAOYNA-N
Synonym ethyl 2-nitropropionate, propanoic acid, 2-nitro-, ethyl ester, propionic acid, 2-nitro-, ethyl ester, ethyl alpha-nitropropionate, acmc-1ce4b, ethyl .alpha.-nitropropionate, propanoicacid, 2-nitro-, ethyl ester
PubChem CID 97978
IUPAC Name ethyl 2-nitropropanoate
SMILES CCOC(=O)C(C)[N+]([O-])=O

Specifications

Density 1.13
Boiling Point 190°C to 195°C
Flash Point 89°C (192°F)
Refractive Index 1.421
Quantity 1 g
Beilstein 774734
Solubility Information Not miscible in water.
Formula Weight 147.13
Percent Purity 96%
Chemical Name or Material Ethyl 2-nitropropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.