missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxo-4-phenylbutyrate, 90+%

Product Code. 11410691
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410691 10 g 10g
11420691 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11410691 Supplier Thermo Scientific Alfa Aesar Supplier No. B20148.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. Se returpolicy

CAS: 64920-29-2 | C12H14O3 | 206.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 64920-29-2
Molecular Formula C12H14O3
Molecular Weight (g/mol) 206.241
MDL Number MFCD00037533
InChI Key STPXIOGYOLJXMZ-UHFFFAOYSA-N
Synonym ethyl 2-oxo-4-phenylbutyrate, ethyl benzylpyruvate, ethyl-2-oxo-4-phenylbutyrate, unii-5ynu9lpt51, 5ynu9lpt51, 2-oxo-4-phenylbutyric acid ethyl ester, 2-oxo-4-phenyl butyric acid ethyl ester, ethyl 2-oxo-4-phenyl-butanoate, 2-oxo-4-phenylbutanoic acid ethyl ester, ethyl2-oxo-4-phenylbutyrate
PubChem CID 562087
IUPAC Name ethyl 2-oxo-4-phenylbutanoate
SMILES CCOC(=O)C(=O)CCC1=CC=CC=C1

Specifikationer

Density 1.09
Boiling Point 131°C to 133°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.502
Quantity 10 g
Beilstein 2725083
Formula Weight 206.24
Percent Purity ≥90%
Chemical Name or Material Ethyl 2-oxo-4-phenylbutyrate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.