missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-oxocyclohexanecarboxylate, 95%

Product Code. 15445838
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445838 100 g 100g
15435838 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445838 Supplier Thermo Scientific Alfa Aesar Supplier No. H59495.22
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1655-07-8 | C9H14O3 | 170.21 g/mol

Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-oxocyclohexanecarboxylate is used to produce other chemicals. For example, it can react with 4-methyl-aniline to get 2-oxo-cyclohexanecarboxylic acid p-toluidide. The reaction occurs with reagent DMAP

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1655-07-8
Molecular Formula C9H14O3
Molecular Weight (g/mol) 170.21
MDL Number MFCD00001631
InChI Key FGSGHBPKHFDJOP-UHFFFAOYNA-N
Synonym ethyl 2-oxocyclohexanecarboxylate, ethyl 2-cyclohexanonecarboxylate, 2-carbethoxycyclohexanone, ethyl cyclohexanone-2-carboxylate, cyclohexanecarboxylic acid, 2-oxo-, ethyl ester, 2-ethoxycarbonyl cyclohexanone, 2-cyclohexanonecarboxylic acid ethyl ester, 2-oxocyclohexanecarboxylic acid ethyl ester, ethyl 2-oxo-1-cyclohexanecarboxylate, ethyl 2-oxocylohexanecarboxylate
PubChem CID 95543
IUPAC Name ethyl 2-oxocyclohexane-1-carboxylate
SMILES CCOC(=O)C1CCCCC1=O

Specifications

Density 1.064
Boiling Point 106°C (11 mmHg)
Flash Point 96°C (205°F)
Linear Formula (O=)C6H9CO2C2H5
Refractive Index 1.4775
Quantity 100 g
Beilstein 608356
Solubility Information Soluble in water.
Formula Weight 170.21
Percent Purity 95%
Chemical Name or Material Ethyl 2-oxocyclohexanecarboxylate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.