missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-pyridylacetate 98%, ACROS Organics™

Product Code. 10498640 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10498640 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10498640 Supplier Thermo Scientific Acros Supplier No. 118910050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2739-98-2
Color Yellow
Density 1.084g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C9H11NO2
Linear Formula (C5H4N)CH2COOC2H5
Refractive Index 1.4963 to 1.4983 (20°C, 589nm)
MDL Number MFCD00006359
Quantity 5 g
Synonym ethyl 2-pyridylacetate, ethyl 2-pyridin-2-yl acetate, ethyl 2-pyridinylacetate, ethyl pyridine-2-acetate, 2-pyridineacetic acid ethyl ester, ethyl 2-pyridineacetate, ethyl 2-2-pyridyl acetate, 2-pyridylacetic acid ethyl ester, ethyl pyridin-2-ylacetate, 2-pyridineacetic acid, ethyl ester
InChI Key IUDKTVXSXWAKJO-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=CC=CC=N1
IUPAC Name ethyl 2-pyridin-2-ylacetate
Molecular Weight (g/mol) 165.192
PubChem CID 75960
Formula Weight 165.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material Ethyl 2-pyridylacetate, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.