missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15417985
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15417985 1 g 1g
15427985 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417985 Supplier Thermo Scientific Alfa Aesar Supplier No. H32065.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306960-67-8
Molecular Formula C9H9F3N2O2
Molecular Weight (g/mol) 234.178
MDL Number MFCD07367386
InChI Key LYEKBQSEKSEXLT-UHFFFAOYSA-N
Synonym ethyl 4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, ethyl 2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate, ethyl-2-trifluoromethyl-4-methyl-5-pyrimidine carboxylate, ethyl4-methyl-2-trifluoromethyl pyrimidine-5-carboxylate, 5-pyrimidinecarboxylicacid, 4-methyl-2-trifluoromethyl-, ethyl ester
PubChem CID 10609709
IUPAC Name ethyl 4-methyl-2-(trifluoromethyl)pyrimidine-5-carboxylate
SMILES CCOC(=O)C1=CN=C(N=C1C)C(F)(F)F

Specifications

Boiling Point 54°C (0.5 mmHg)
Quantity 1 g
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material Ethyl 2-trifluoromethyl-4-methylpyrimidine-5-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.