missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-[2,6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate, 97%, Thermo Scientific™

Product Code. 10163201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10163201 1 g Glass bottle 1g
10757592 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10163201

Brand: Thermo Scientific Acros 335410010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96568-04-6
Molecular Formula C10H8Cl2FNO3
Molecular Weight (g/mol) 280.08
MDL Number MFCD00799535
InChI Key IEUHWNLWVMLHHC-UHFFFAOYSA-N
Synonym ethyl 3-2,6-dichloro-5-fluoropyridin-3-yl-3-oxopropanoate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridyl-3-oxopropionate, ethyl 2,6-dichloro-5-fluoronicotinoylacetate, ethyl 2,6-dichloro-5-fluoro pyridine-3-acetoacetate, ethyl 2,6-dichloro-5-fluoronicotinoyl acetate, ethyl 2,6-dichloro-5-fluornicotinoylacetate, ethyl 3-2,6-dichloro-5-fluoro-3-pyridiyl-3-oxopropanoate, 2,6-dichloro-5-fluoronicotinoyl acetic acid ethyl ester, ethyl 2,6-dichloro-5-fluoro nicotinylacetate, ethyl 3-2,6-dichloro-5-fluoro 3-pyridyl-3-oxopropanoate
PubChem CID 2733708
IUPAC Name ethyl 3-(2,6-dichloro-5-fluoropyridin-3-yl)-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1=CC(F)=C(Cl)N=C1Cl

Specifications

Melting Point 68°C to 72°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 280.08
Percent Purity 97%
Chemical Name or Material Ethyl 3-[2, 6-dichloro-5-fluoro-(3-pyridiyl)]-3-oxopropanoate, 97%
compliance-icons
Product Identifier
  • Ethyl 3-¢2,6-dichloro-5-fluoro-(3-pyridiyl)!-3-oxopropanoate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.