missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-furyl)-1-methyl-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10143314
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143314 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10143314 Supplier Thermo Scientific Maybridge Supplier No. CC47023DA
 Restricted Item
Estimated Shipment: 19-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 104295-62-7
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H12N2O3
MDL Number MFCD08271951
Quantity 1 g
Synonym ethyl 3-2-furyl-1-methyl-1h-pyrazole-5-carboxylate, ethyl 5-furan-2-yl-2-methylpyrazole-3-carboxylate, 5-ethoxycarbonyl-3-2-furyl-1-methylpyrazole, 1h-pyrazole-5-carboxylicacid, 3-2-furanyl-1-methyl-, ethyl ester, acmc-1bsv1, ethyl 3-2-furyl-1-methylpyrazole-5-carboxylate, ethyl 3-furan-2-yl-1-methyl-1h-pyrazole-5-carboxylate, 1-methyl-3-2-furyl-1h-pyrazole-5-carboxylic acid ethyl ester, 1h-pyrazole-5-carboxylicacid,3-2-furanyl-1-methyl-,ethyl ester
InChI Key RMIOHJSPVQALON-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC(=NN1C)C2=CC=CO2
IUPAC Name ethyl 5-(furan-2-yl)-2-methylpyrazole-3-carboxylate
Molecular Weight (g/mol) 220.228
PubChem CID 7537645
Formula Weight 220.23
Percent Purity 97%
Chemical Name or Material Ethyl 3-(2-furyl)-1-methyl-1H-pyrazole-5-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.