missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-thienyl)-1H-pyrazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10772323
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10772323 1 g Amber Glass Bottle 1g
10437184 5 g Amber Glass Bottle 5g
10455142 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10772323 Supplier Thermo Scientific Maybridge Supplier No. SPB04031DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 121195-03-7
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.262
MDL Number MFCD07772880
InChI Key NTCRDZSJYGZRIE-UHFFFAOYSA-N
Synonym ethyl 5-thien-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-thiophen-2-yl-1h-pyrazole-5-carboxylate, ethyl 5-2-thienyl pyrazole-3-carboxylate, ethyl 5-thiophen-2-yl-1h-pyrazole-3-carboxylate, ethyl 3-2-thienyl-1h-pyrazole-5-carboxylate, ethyl 5-thiophen-2-yl-2h-pyrazole-3-carboxylate, 1h-pyrazole-3-carboxylic acid, 5-2-thienyl-, ethyl ester, 1h-pyrazole-3-carboxylicacid, 5-2-thienyl-, ethyl ester, acmc-20mpcx, maybridge1_008811
PubChem CID 2736478
IUPAC Name ethyl 5-thiophen-2-yl-1H-pyrazole-3-carboxylate
SMILES CCOC(=O)C1=NNC(=C1)C2=CC=CS2

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.27
Percent Purity 95%
Chemical Name or Material Ethyl 5-thien-2-yl-1H-pyrazole-3-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.