missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(2-thienyl)piperazine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 15407595
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15407595 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15407595 Supplier Thermo Scientific Alfa Aesar Supplier No. H26527.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 85803-50-5
Molecular Formula C11H16N2O2S
Refractive Index 1.5485
MDL Number MFCD06245431
Quantity 250 mg
Synonym ethyl 3-thiophen-2-yl piperazine-1-carboxylate, ethyl 3-2-thienyl piperazine-1-carboxylate, 3-thiophen-2-yl-piperazine-1-carboxylic acid ethyl ester, acmc-20amyi, ethyl 3-2-thienyl piperazinecarboxylate, ethyl 3-2-thienyl piperazine-carboxylate, 3-thien-2-yl-piperazine-1-carboxylic acid ethyl ester, 3-thiophen-2-yl-piperazine-1-carboxylic acidethyl ester, 3-thiophen-2-ylpiperazine-1-carboxylic acid ethyl ester, ethyl 3-thiophen-2-yl-piperazine-1-carboxylate
InChI Key YNIWHEQLSSKQCX-UHFFFAOYSA-N
SMILES CCOC(=O)N1CCNC(C1)C2=CC=CS2
IUPAC Name ethyl 3-thiophen-2-ylpiperazine-1-carboxylate
Molecular Weight (g/mol) 240.321
PubChem CID 2771741
Formula Weight 240.33
Percent Purity 95%
Chemical Name or Material Ethyl 3-(2-thienyl)piperazine-1-carboxylate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.