missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3-diethoxypropanoate, 98%

Product Code. 10083330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10083330 5 mL Glass bottle 5mL
10023360 25 mL Glass bottle 25mL
10521984 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10083330 Supplier Thermo Scientific Acros Supplier No. 269640050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10601-80-6 | C9H18O4 | 190.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10601-80-6
Molecular Formula C9H18O4
Molecular Weight (g/mol) 190.24
MDL Number MFCD00009865
InChI Key SIALOQYKFQEKOG-UHFFFAOYSA-N
Synonym ethyl 3,3-diethoxypropionate, 3,3-diethoxypropionic acid ethyl ester, ethyl3,3-diethoxypropionate, propanoic acid, 3,3-diethoxy-, ethyl ester, 3,3-diethoxy-propionic acid ethyl ester, malonaldehydic acid ethyl ester diethylacetal, zlchem 288, pubchem18874, acmc-1bujf, ethyl 3.3-diethoxypropanoate
PubChem CID 66389
IUPAC Name ethyl 3,3-diethoxypropanoate
SMILES CCOC(CC(=O)OCC)OCC

Specifications

Color Colorless
Density 0.9700g/mL
Boiling Point 95°C (17.0 mmHg)
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5O)2CHCH2COOC2H5
Refractive Index 1.4100 to 1.412
Quantity 5 mL
Specific Gravity 0.97
Solubility Information Solubility in water: 23g/L (20 c)
Formula Weight 190.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 3, 3-diethoxypropanoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.