missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(4-fluoroanilino)crotonate, 97%, Thermo Scientific™

Product Code. 11453787
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453787 25 g 25g
11443787 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11453787 Supplier Thermo Scientific Alfa Aesar Supplier No. A18251.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18529-17-4
Molecular Formula C12H14FNO2
Molecular Weight (g/mol) 223.247
MDL Number MFCD00016720
InChI Key BUDOLSNUNUYBLF-HJWRWDBZSA-N
Synonym ethyl 3-4-fluoroanilino crotonate, z-ethyl 3-4-fluorophenyl amino but-2-enoate, ethyl 2z-3-4-fluorophenyl amino but-2-enoate, 3-4-fluorophenylamino but-2-enoic acid ethyl ester, ethyl z-3-4-fluoroanilino but-2-enoate, z-3-4-fluorophenylamino-2-butenoic acid ethyl ester
PubChem CID 5811492
IUPAC Name ethyl (Z)-3-(4-fluoroanilino)but-2-enoate
SMILES CCOC(=O)C=C(C)NC1=CC=C(C=C1)F

Specifications

Quantity 25 g
Formula Weight 223.25
Percent Purity 97%
Chemical Name or Material Ethyl 3-(4-fluoroanilino)crotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.