missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-amino-4,4,4-trifluorocrotonate, 97%

Product Code. 11495107
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495107 25 g 25g
11485107 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495107 Supplier Thermo Scientific Alfa Aesar Supplier No. B22545.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-29-2 | C6H8F3NO2 | 183.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-29-2
Molecular Formula C6H8F3NO2
Molecular Weight (g/mol) 183.13
MDL Number MFCD00068195
InChI Key NXVKRKUGIINGHD-ONEGZZNKSA-N
PubChem CID 10899311
IUPAC Name ethyl (E)-3-amino-4,4,4-trifluorobut-2-enoate
SMILES CCOC(=O)C=C(C(F)(F)F)N

Specifications

Melting Point 24°C to 26°C
Density 1.245
Boiling Point 83°C to 85°C (15 mmHg)
Flash Point 65°C (149°F)
Odor Amine-like
Refractive Index 1.4229
Quantity 25 g
UN Number UN3259
Beilstein 4397839
Sensitivity Air Sensitive
Synonym ethyl 3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluorobut-2-enoate, ethyl 2z-3-amino-4,4,4-trifluorobut-2-enoate, 3-amino-4,4,4-trifluorocrotonic acid ethyl ester, ethyl-3-amino-4,4,4-trifluorocrotonate, ethyl 3-amino-4,4,4-trifluoro-2-butenoate, ethyl z-3-amino-4,4,4-trifluorobut-2-enoate, ethyl z-3-amino-4,4,4-trifluoro-but-2-enoate, hcvdpjfpdddldlbdlrfuulu@bh, ethyl3-amino-4,4,4-trifluorocrotonate
Formula Weight 183.13
Percent Purity 97%
Chemical Name or Material Ethyl 3-amino-4,4,4-trifluorocrotonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.