missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-aminocrotonate, 98+%

Product Code. 11419873
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419873 25 g 25g
11439873 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419873 Supplier Thermo Scientific Alfa Aesar Supplier No. A12991.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 626-34-6 | C6H11NO2 | 129.16 g/mol

Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-aminocrotonate, is used as an intermediate in organic synthesis. It is also used as an active ingredient in pharmaceutical synthesis and feed auxiliary agent. It is also used as an active pharmaceutical intermediate.

Solubility
Soluble in water (26 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible to acids, bases, oxidizing agents, reducing agents. Air & Moisture Sensitive & Hygroscopic.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-34-6
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
MDL Number MFCD00008073,MFCD02730138
InChI Key YPMPTULBFPFSEQ-UHFFFAOYSA-N
Synonym z-ethyl 3-aminobut-2-enoate, ethyl 3-aminocrotonate, ethyl 3-aminobut-2-enoate, ethyl 2z-3-aminobut-2-enoate, 2-butenoic acid, 3-amino-, ethyl ester, 2z, ethyl z-3-aminobut-2-enoate, ethyl 3-amino-2-butenoate, .beta.-aminocrotonic acid ethyl ester, 2-butenoic acid, 3-amino-, ethyl ester, ethyl .beta.-aminocrotonate
PubChem CID 643756
IUPAC Name ethyl (Z)-3-aminobut-2-enoate
SMILES CCOC(=O)C=C(C)N

Specifications

Melting Point 29°C to 35°C
Boiling Point 210°C to 215°C
Flash Point 97°C (206°F)
Quantity 25 g
UN Number UN3263
Beilstein 471284
Formula Weight 129.16
Percent Purity ≥98%
Chemical Name or Material Ethyl 3-aminocrotonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.