missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Aminocrotonate, TRC

Product Code. 30375852
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30375852 25 g
30373761 100 g
30372916 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30375852 Supplier Toronto Research Chemicals Supplier No. TRCE89808025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7318-00-5
Molecular Formula C6H11NO2
Molecular Weight (g/mol) 129.16
Synonym 3-Amino-2-butenoic Acid Ethyl Ester, beta-Aminocrotonic Acid Ethyl Ester, 3-Aminocrotonic Acid Ethyl Ester, Ethyl 3-Amino-2-butenoate, Ethyl beta-Amino-beta-methylacrylate, Ethyl beta-Aminocrotonate, NSC 1086, beta-Aminocrotonic Acid Ethyl Ester
SMILES CCOC(=O)C=C(C)N

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H11NO2/c1-3-9-6(8)4-5(2)7/h4H,3,7H2,1-2H3/b5-4+
Formula Weight 129.079
Chemical Name or Material Ethyl 3-Aminocrotonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.