missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate, 96%, Thermo Scientific Chemicals

Product Code. 15488715
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15488715 1 g 1g
15498715 5 g 5g
15408725 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15488715 Supplier Thermo Scientific Alfa Aesar Supplier No. H33967.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 128073-16-5 | Molecular Formula: C9H7ClF3NO2 | Molecular Weight: 253.605 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 128073-16-5
Molecular Formula C9H7ClF3NO2
Molecular Weight (g/mol) 253.605
MDL Number MFCD06656414
InChI Key COMQYNZHBCNPNW-UHFFFAOYSA-N
Synonym ethyl 3-chloro-5-trifluoromethyl picolinate, ethyl 3-chloro-5-trifluoromethyl pyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl-2-pyridinecarboxylate, 2-pyridinecarboxylic acid, 3-chloro-5-trifluoromethyl-, ethyl ester, 3-chloro-5-trifluoromethylpyridine-2-carboxylic acid ethyl ester, ethyl 5-trifluoromethyl-3-chloropyridine-2-carboxylate, ethyl 3-chloro-5-trifluoromethyl picolite, ethyl 3-chloro-5trifluoromethyl-2-pyridinecarboxylate
PubChem CID 22013558
IUPAC Name ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate
SMILES CCOC(=O)C1=C(C=C(C=N1)C(F)(F)F)Cl

Specifica

Boiling Point 64°C to 66°C (0.008 mmHg)
Quantity 1 g
Formula Weight 253.6
Percent Purity 96%
Chemical Name or Material Ethyl 3-chloro-5-(trifluoromethyl)pyridine-2-carboxylate

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.