missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-chloropropionate, 98%

Product Code. 11934931 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11934931 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11934931 Supplier Thermo Scientific Acros Supplier No. 118261000
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-71-2 | C5H9ClO2 | 136.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 623-71-2
Density 1.0030g/mL
Boiling Point 162.0°C to 163.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H9ClO2
Linear Formula ClCH2CH2CO2C2H5
Refractive Index 1.4240 to 1.4260
MDL Number MFCD00000989
Quantity 100 g
Beilstein 02,25
Specific Gravity 1.003
Synonym ethyl 3-chloropropionate, 3-chloropropionic acid ethyl ester, propanoic acid, 3-chloro-, ethyl ester, ethyl beta-chloropropionate, propionic acid, 3-chloro-, ethyl ester, ethyl .beta.-chloropropionate, 3-chloropropionicacidethylester, 3-chloro-propanoicacidethylester, ethyl-3-chlorpropanoat, ethyl3-chloropropionate
Solubility Information Solubility in water: insoluble.
InChI Key ZCLGVXACCAZJOX-UHFFFAOYSA-N
SMILES CCOC(=O)CCCl
IUPAC Name ethyl 3-chloropropanoate
Molecular Weight (g/mol) 136.58
PubChem CID 69341
Formula Weight 136.58
Percent Purity 98%
Chemical Name or Material Ethyl 3-chloropropionate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl 3-chloropropionate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.