missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyclopropyl-3-oxopropanoate, TRC

Product Code. 30382944
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382944 10 g -
30385291 50 g -
30371275 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382944 Supplier Toronto Research Chemicals Supplier No. TRCE92632810G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 24922-02-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
IUPAC Name ethyl 3-cyclopropyl-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1CC1

Especificaciones

Quantity 10 g
InChI Formula InChI=1S/C8H12O3/c1-2-11-8(10)5-7(9)6-3-4-6/h6H,2-5H2,1H3
Formula Weight 156.0786
Chemical Name or Material Ethyl 3-cyclopropyl-3-oxopropanoate
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.