missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyclopropyl-3-oxopropanoate, TRC

Product Code. 30385291
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30385291 50 g -
30382944 10 g -
30371275 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30385291 Supplier Toronto Research Chemicals Supplier No. TRCE92632850G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24922-02-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
IUPAC Name ethyl 3-cyclopropyl-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1CC1

Specifications

Quantity 50 g
InChI Formula InChI=1S/C8H12O3/c1-2-11-8(10)5-7(9)6-3-4-6/h6H,2-5H2,1H3
Formula Weight 156.0786
Chemical Name or Material Ethyl 3-cyclopropyl-3-oxopropanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.