missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-cyclopropyl-3-oxopropionate, 96%

Product Code. 15416617
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15416617 5 g 5g
15406617 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15416617 Supplier Thermo Scientific Alfa Aesar Supplier No. H33756.06
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 24922-02-9 | C8H12O3 | 156.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24922-02-9
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.18
MDL Number MFCD00184570
InChI Key LFSVADABIDBSBV-UHFFFAOYSA-N
Synonym 3-cyclopropyl-3-oxo-propionic acid ethyl ester, ethyl 3-cyclopropyl-3-oxo-propanoate, ethyl 3-cyclopropyl-3-oxopropionate, ethyl 3-cyclopropyl-3-oxo-propionate, ethyl beta-oxo-cyclopropane propionate, ksc205o7f, ethyl cyclopropylcarbonylacetate, carbethoxymethyl cyclopropyl ketone, ethyl ?-oxo-cyclopropanepropionate, ethyl3-cyclopropyl-3-oxopropanoate
PubChem CID 262979
IUPAC Name ethyl 3-cyclopropyl-3-oxopropanoate
SMILES CCOC(=O)CC(=O)C1CC1

Specifications

Boiling Point 73°C (1 mmHg)
Quantity 5 g
Formula Weight 156.18
Percent Purity 97%
Chemical Name or Material Ethyl 3-cyclopropyl-3-oxopropionate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.