missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(dimethylamino)acrylate, 99%

Product Code. 11415707
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415707 10 g 10g
11425707 50 g 50g
11435707 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415707 Supplier Thermo Scientific Alfa Aesar Supplier No. B24291.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 924-99-2 | C7H13NO2 | 143.186 g/mol

Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-(dimethylamino)acrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water 90 g/L (25°C).

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-99-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.186
MDL Number MFCD00144269
InChI Key MVUMJYQUKKUOHO-AATRIKPKSA-N
Synonym ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
PubChem CID 5369162
IUPAC Name ethyl (E)-3-(dimethylamino)prop-2-enoate
SMILES CCOC(=O)C=CN(C)C

Specifications

Melting Point 17°C to 18°C
Boiling Point 118°C to 121°C (10 mmHg)
Linear Formula (CH3)2NCH=CHCO2CH2CH3
Refractive Index 1.512
Quantity 10 g
Solubility Information Soluble in water 90g/L (25°C).
Formula Weight 143.19
Percent Purity 99%
Chemical Name or Material Ethyl 3-(dimethylamino)acrylate
compliance-icons
Product Identifier
  • Ethyl 3-(dimethylamino)acrylate
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Ethyl trans-3-dimethylaminoacrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.