missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-hydroxyhexanoate, 97%

Product Code. 11429952
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429952 100 g 100g
11419952 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11429952 Supplier Thermo Scientific Alfa Aesar Supplier No. A12060.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2305-25-1 | C8H16O3 | 160.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2305-25-1
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00036604
InChI Key LYRIITRHDCNUHV-UHFFFAOYSA-N
Synonym hexanoic acid, 3-hydroxy-, ethyl ester, ethyl beta-hydroxycaproate, ethyl 3-hydroxy-hexanoate, ethyl hydroxy-3-hexanoate, 3-hydroxyhexanoic acid ethyl ester, fema no. 3545, acmc-20lakf, ethyl 3-hydroxycaproate, ethyl-3-hydrixyhexanoate, ethyl 3-hydroxy hexanoate
PubChem CID 61293
ChEBI CHEBI:23997
IUPAC Name ethyl 3-hydroxyhexanoate
SMILES CCCC(CC(=O)OCC)O

Specifications

Density 0.974
Boiling Point 90°C to 92°C (14 mmHg)
Flash Point 94°C (202°F)
Linear Formula CH3CH2CH2CHOHCH2CO2CH2CH3
Refractive Index 1.4298
Quantity 100 g
Formula Weight 160.21
Percent Purity 97%
Chemical Name or Material Ethyl 3-hydroxyhexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.