missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-indoleacetate, 99%, Thermo Scientific™

Product Code. 10682522 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682522 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10682522 Supplier Thermo Scientific Acros Supplier No. 331570050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 778-82-5
Melting Point 42.0°C to 45.0°C
Color White
Boiling Point 164.0°C to 166.0°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C12H13NO2
MDL Number MFCD00005635
Quantity 5 g
Synonym ethyl 3-indoleacetate, ethyl 2-1h-indol-3-yl acetate, indole-3-acetic acid ethyl ester, ethyl indole-3-acetate, ethyl indol-3-ylacetate, 1h-indole-3-acetic acid, ethyl ester, ethyl 1h-indol-3-ylacetate, indole-3-acetic acid, ethyl ester, ethyl beta-indolylacetate, ethyl3-indoleacetate
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone
InChI Key HUDBDWIQSIGUDI-UHFFFAOYSA-N
SMILES CCOC(=O)CC1=CNC2=CC=CC=C21
IUPAC Name ethyl 2-(1H-indol-3-yl)acetate
Molecular Weight (g/mol) 203.24
PubChem CID 13067
Formula Weight 203.24
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 3-indoleacetate, 99%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.