missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isocyanatopropionate, 98%

Product Code. 11332167
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332167 5 g 5g
11322167 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332167 Supplier Thermo Scientific Alfa Aesar Supplier No. L09616.06
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5100-34-5 | C6H9NO3 | 143.14 g/mol

Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.

Solubility
Not miscible with water.

Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5100-34-5
Molecular Formula C6H9NO3
Molecular Weight (g/mol) 143.14
MDL Number MFCD00078461
InChI Key XBSGYVHOINMTIM-UHFFFAOYSA-N
Synonym ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
PubChem CID 2733373
IUPAC Name ethyl 3-isocyanatopropanoate
SMILES CCOC(=O)CCN=C=O

Specifications

Density 1.087
Boiling Point 74°C to 75°C (4 mmHg)
Flash Point 92°C (197°F)
Odor Irritating
Refractive Index 1.429
Quantity 5 g
UN Number UN2206
Beilstein 1705618
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 143.14
Percent Purity 98%
Chemical Name or Material Ethyl 3-isocyanatopropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.