missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-isothiocyanatopropionate, 97%

Product Code. 11373066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11373066 1 g 1g
11383066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373066 Supplier Thermo Scientific Alfa Aesar Supplier No. L10911.03
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 17126-62-4 | C6H9NO2S | 159.203 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17126-62-4
Molecular Formula C6H9NO2S
Molecular Weight (g/mol) 159.203
MDL Number MFCD00041130
InChI Key UPTRONYNXNYITM-UHFFFAOYSA-N
Synonym ethyl 3-isothiocyanatopropionate, 3-isothiocyanato-propionic acid ethyl ester, acmc-1ca2j, ethyl-3-isothiocynato propionate, isothiocyanato-propionic acid ethyl ester, 3-isothiocyanatopropionic acid ethyl ester, propanoic acid,3-isothiocyanato-,ethyl ester, propanoic acid,3-isothiocyanato-, ethyl ester
PubChem CID 526514
IUPAC Name ethyl 3-isothiocyanatopropanoate
SMILES CCOC(=O)CCN=C=S

Specifications

Density 1.12
Boiling Point 75°C to 76°C (8 mmHg)
Odor Pungent
Linear Formula SCNCH2CH2CO2CH2CH3
Refractive Index 1.5014
Quantity 1 g
UN Number UN2922
Beilstein 1705633
Sensitivity Moisture sensitive
Formula Weight 159.2
Percent Purity 97%
Chemical Name or Material Ethyl 3-isothiocyanatopropionate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.