missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methylisoxazole-4-carboxylate, 97%

Product Code. 11389125
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389125 1 g 1g
11399125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389125 Supplier Thermo Scientific Alfa Aesar Supplier No. L16362.03
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 20328-15-8 | C7H9NO3 | 155.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20328-15-8
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD01075736
InChI Key AEWHICNMVQOWJE-UHFFFAOYSA-N
Synonym ethyl 3-methylisoxazole-4-carboxylate, ethyl 3-methyl-isoxazole-4-carboxylate, 4-isoxazolecarboxylic acid, 3-methyl-, ethyl ester, acmc-20akze, ethyl 3-methyl-4-isoxazolecarboxylate, 3-methyl-4-isoxazolecarboxylic acid ethyl ester, 3-methyl-isoxazole-4-carboxylic acid ethyl ester, 4-isoxazolecarboxylicacid, 3-methyl-, ethyl ester, 3-methyl-4-isoxazolecarboxylic acid, ethyl ester, 3-methylisoxazole-4-carboxylic acid ethyl ester
PubChem CID 317668
IUPAC Name ethyl 3-methyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=CON=C1C

Specifications

Boiling Point 64°C (0.6 mmHg)
Refractive Index 1.4595
Quantity 1 g
Beilstein 1072756
Formula Weight 155.15
Percent Purity 97%
Chemical Name or Material Ethyl 3-methylisoxazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.