missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(N,N-dimethylamino)acrylate, 99+%

Product Code. 11965341 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11965341 100 g Glass bottle 100g
10790941 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11965341 Supplier Thermo Scientific Acros Supplier No. 269631000
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 924-99-2 | C7H13NO2 | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924-99-2
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00144269
InChI Key MVUMJYQUKKUOHO-AATRIKPKSA-N
Synonym ethyl 3-dimethylamino acrylate, e-ethyl 3-dimethylamino acrylate, ethyl 3-n,n-dimethylamino acrylate, ethyl n,n-dimethylaminoacrylate, ethyl trans-3-dimethylaminoacrylate, ethyl 2e-3-dimethylamino prop-2-enoate, ethyl e-3-dimethylamino prop-2-enoate, ethyl 3-dimethylamino prop-2-enoate, acrylic acid, 3-dimethylamino-, ethyl ester, e, 2-propenoic acid, 3-dimethylamino-, ethyl ester, 2e
PubChem CID 5369162
IUPAC Name ethyl (E)-3-(dimethylamino)prop-2-enoate
SMILES CCOC(=O)C=CN(C)C

Specifications

Melting Point 17.0°C to 18.0°C
Color Orange to Yellow
Density 1.0000g/mL
Boiling Point 118°C to 121°C (10.0 HPA)
Flash Point 105°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH=CHCOOC2H5
Refractive Index 1.5105 to 1.5125
Quantity 100 g
Specific Gravity 1
Solubility Information Solubility in water: 90g/L (25 c)
Formula Weight 143.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Ethyl 3-(N, N-dimethylamino)acrylate, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl 3-(N,N-dimethylamino)acrylate
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.