missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenylglycidate, 90%, mixture of cis and trans

Product Code. 10595041 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
5mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10595041 5 mL Glass bottle 5mL
10327711 50 mL Glass bottle 50mL
10051761 250 mL Glass bottle 250mL
10647884 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10595041 Supplier Thermo Scientific Acros Supplier No. 332650050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 121-39-1 | C11H12O3 | 192.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-39-1
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
MDL Number MFCD00005123
InChI Key GOMAKLPNAAZVCJ-UHFFFAOYSA-N
PubChem CID 8469
IUPAC Name ethyl 3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)C2=CC=CC=C2

Specifications

Color Colorless to Yellow
Density 1.1200g/mL
Boiling Point 96.0°C (0.5 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5165 to 1.5205
Quantity 5 mL
Beilstein 18,42
Specific Gravity 1.12
Synonym ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
Solubility Information Solubility in water: insoluble
Formula Weight 192.21
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Ethyl 3-phenylglycidate, 90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.