missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-phenylglycidate, cis + trans, 90+%

Product Code. 11476156
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476156 50 mL 50mL
11486156 250 mL 250mL
11466156 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11476156 Supplier Thermo Scientific Alfa Aesar Supplier No. B22990.AD
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 121-39-1 | C11H12O3 | 192.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-39-1
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00005123
InChI Key GOMAKLPNAAZVCJ-UHFFFAOYSA-N
PubChem CID 8469
IUPAC Name ethyl 3-phenyloxirane-2-carboxylate
SMILES CCOC(=O)C1C(O1)C2=CC=CC=C2

Specifications

Density 1.102
Boiling Point 95°C to 97°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.518
Quantity 50 mL
Sensitivity Moisture sensitive
Synonym ethyl 3-phenylglycidate, ethyl phenylglycidate, oxiranecarboxylic acid, 3-phenyl-, ethyl ester, ethyl 2,3-epoxy-3-phenylpropionate, ethyl 3-phenyloxiranecarboxylate, glycidic acid, 3-phenyl-, ethyl ester, 2-ethyl-3-phenylglycidate, fema no. 2454, ccris 2623, ethyl3-phenylglycidate
Formula Weight 192.21
Percent Purity ≥90%
Chemical Name or Material Ethyl 3-phenylglycidate, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.