missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Phenylpropionate, TRC

Product Code. 30384259
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384259 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384259 Supplier Toronto Research Chemicals Supplier No. TRCE925720100G
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrocinnamic acid ethyl ester
CAS 2021-28-5
Molecular Formula C11 H14 O2
Quantity 100 g
Synonym Benzenepropanoic acid, ethyl ester, Hydrocinnamic acid, ethyl ester (6CI,8CI), 3-Phenylpropanoic acid ethyl ester, 3-Phenylpropionic acid ethyl ester, Ethyl 3-phenylpropanoate, Ethyl 3-phenylpropionate, Ethyl benzenepropanoate, Ethyl dihydrocinnamate, Ethyl hydrocinnamate, NSC 126040
InChI Formula InChI=1S/C11H14O2/c1-2-13-11(12)9-8-10-6-4-3-5-7-10/h3-7H,2,8-9H2,1H3
SMILES CCOC(=O)CCc1ccccc1
IUPAC Name ethyl 3-phenylpropanoate
Molecular Weight (g/mol) 178.23
Formula Weight 178.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.