missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(tert-butyl)-1-methyl-1H-pyrazole-5-carboxylate, Thermo Scientific™

Product Code. 10580933
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10580933 10 g 10g
10110662 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10580933 Supplier Thermo Scientific Maybridge Supplier No. SPB03148EA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 133261-10-6
Molecular Formula C11H18N2O2
Molecular Weight (g/mol) 210.277
InChI Key JCOHRMJIHAFFBC-UHFFFAOYSA-N
PubChem CID 2799344
IUPAC Name ethyl 5-tert-butyl-2-methylpyrazole-3-carboxylate
SMILES CCOC(=O)C1=CC(=NN1C)C(C)(C)C

Specifications

Quantity 10 g
Synonym ethyl 3-tert-butyl-1-methyl-1h-pyrazole-5-carboxylate, ethyl 3-tert-butyl-1-methylpyrazole-5-carboxylate, 1h-pyrazole-5-carboxylicacid, 3-1,1-dimethylethyl-1-methyl-, ethyl ester, acmc-1c1ez, ethyl3-tert-butyl-1-methylpyrazole-5-carboxylate, 5-tert-butyl-2-methyl-pyrazole-3-carboxylic acid ethyl ester, 5-tert-butyl-2-methyl-2h-pyrazole-3-carboxylic acid ethyl ester
Formula Weight 210
Chemical Name or Material Ethyl 3-(tert-butyl)-1-methyl-1H-pyrazole-5-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.