missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate, 97%

Product Code. 11490164
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490164 25 g 25g
11470164 1 g 1g
11480164 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490164 Supplier Thermo Scientific Alfa Aesar Supplier No. A13609.14
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 155377-19-8 | C7H7F3N2O2 | 208.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 155377-19-8
Molecular Formula C7H7F3N2O2
Molecular Weight (g/mol) 208.14
MDL Number MFCD00052083
InChI Key VYXIHSAEOXPAEY-UHFFFAOYSA-N
Synonym ethyl 3-trifluoromethyl pyrazole-4-carboxylate, ethyl 3-trifluoromethyl-1h-pyrazole-4-carboxylate, ethyl 5-trifluoromethyl-1h-pyrazole-4-carboxylate, 3-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-trifluoromethyl pyrazole-4-carboxylic acid ethyl ester, ethyl 3-trifluoromethyl-2h-pyrazole-4-carboxylate, 5-trifluoromethyl-1h-pyrazole-4-carboxylic acid ethyl ester, 1h-pyrazole-4-carboxylic acid, 3-trifluoromethyl-, ethyl ester
PubChem CID 596095
IUPAC Name ethyl 5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1)C(F)(F)F

Specifications

Melting Point 142°C to 146°C
Quantity 25 g
Formula Weight 208.14
Percent Purity 97%
Chemical Name or Material Ethyl 3-trifluoromethyl-1H-pyrazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.