missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z), 96%

Product Code. 11392207
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392207 10 g 10g
11382207 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11392207 Supplier Thermo Scientific Alfa Aesar Supplier No. L09688.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24490-03-7 | C7H9F3O2 | 182.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24490-03-7
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.14
MDL Number MFCD00040846
InChI Key OSZLARYVWBUKTG-SNAWJCMRSA-N
Synonym ethyl 4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, ethyl 2e-4,4,4-trifluoro-3-methylbut-2-enoate, ethyl 3-trifluoromethyl crotonate, cis + trans, ethyl e-4,4,4-trifluoro-3-methylbut-2-enoate, e-3-trifluoromethyl-2-butenoic acid ethyl ester
PubChem CID 5838607
IUPAC Name ethyl (E)-4,4,4-trifluoro-3-methylbut-2-enoate
SMILES CCOC(=O)\C=C(/C)C(F)(F)F

Specifications

Boiling Point 129°C to 130°C
Flash Point 32°C (89°F)
Refractive Index 1.378
Quantity 10 g
UN Number UN3272
Beilstein 1910081
Formula Weight 182.14
Percent Purity 96%
Chemical Name or Material Ethyl 3-(trifluoromethyl)crotonate, (E)+(Z)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.