missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 97%, Thermo Scientific Chemicals

Product Code. 11384356
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384356 5 g 5g
11374356 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384356 Supplier Thermo Scientific Alfa Aesar Supplier No. L19969.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91600-33-8
Molecular Formula C7H11F3O3
Molecular Weight (g/mol) 200.157
MDL Number MFCD00190634
InChI Key MFRIOKNLYRUYHP-UHFFFAOYSA-N
Synonym ethyl 2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutyrate, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutanoate, ethyl-4,4,4-trifluor-3-hydroxy-2-methylbutanoat, ethyl 2-methyl-4,4,4-trifluoro-3-hydroxybutyrate, ethyl-2-methyl-3-hydroxy-4,4,4-trifluorobutyrate, ethyl 3-hydroxy-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate, 4,4,4-trifluoro-3-hydroxy-2-methylbutyric acid ethyl ester
PubChem CID 533787
IUPAC Name ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutanoate
SMILES CCOC(=O)C(C)C(C(F)(F)F)O

Specifications

Boiling Point 180°C to 184°C
Quantity 5 g
Beilstein 6194166
Formula Weight 200.16
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.