missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluoro-3-hydroxybutyrate, 97%, Thermo Scientific™

Product Code. 11313356
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11313356 50 g 50g
11303356 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11313356 Supplier Thermo Scientific Alfa Aesar Supplier No. L12753.18
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-30-5
Molecular Formula C6H9F3O3
Molecular Weight (g/mol) 186.13
MDL Number MFCD00040842
InChI Key ZWEDFBKLJILTMC-UHFFFAOYNA-N
Synonym ethyl 3-hydroxy-4,4,4-trifluorobutyrate, ethyl 4,4,4-trifluoro-3-hydroxybutyrate, ethyl 3-hydroxy-4,4,4-trifluorobutanoate, 3-hydroxy-4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-3-hydroxybutyric acid ethyl ester, ethyl3-hydroxy-4,4,4-trifluorobutyrate, butanoic acid, 4,4,4-trifluoro-3-hydroxy-, ethyl ester, acmc-209irs, acmc-20al28, ksc496k8b
PubChem CID 136210
IUPAC Name ethyl 4,4,4-trifluoro-3-hydroxybutanoate
SMILES CCOC(=O)CC(O)C(F)(F)F

Specifications

Melting Point 23°C
Boiling Point 90°C to 91°C (50 mmHg)
Refractive Index 1.3745
Quantity 50 g
Beilstein 1708105
Formula Weight 186.13
Percent Purity 97%
Chemical Name or Material Ethyl 4,4,4-trifluoro-3-hydroxybutyrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.