missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluorobutyrate, 98%, Thermo Scientific™

Product Code. 10077492 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10077492 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10077492 Supplier Thermo Scientific Acros Supplier No. 309650050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 371-26-6
Color Colorless to Yellow
Density 1.1600g/mL
Boiling Point 127.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H9F3O2
Linear Formula CF3CH2CH2CO2C2H5
Refractive Index 1.3510 to 1.3530
MDL Number MFCD00041398
Quantity 5 g
Specific Gravity 1.16
Synonym ethyl 4,4,4-trifluorobutyrate, ethyl4,4,4-trifluorobutyrate, 4,4,4-trifluorobutyric acid ethyl ester, 4,4,4-trifluoro-butyric acid ethyl ester, butanoic acid, 4,4,4-trifluoro-, ethyl ester, acmc-1cncd, ethyl-4,4,4-trifluorobutyrate, ethyl 3-trifluoromethylpropionate, butanoic acid,4,4,4-trifluoro-, ethyl ester
InChI Key PSRZMXNNQTWAGB-UHFFFAOYSA-N
SMILES CCOC(=O)CCC(F)(F)F
IUPAC Name ethyl 4,4,4-trifluorobutanoate
Molecular Weight (g/mol) 170.13
PubChem CID 2733273
Formula Weight 170.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-trifluorobutyrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.