missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,4,4-trifluorocrotonate, 98%, Thermo Scientific™

Product Code. 10125800 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Glass bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10125800 Glass bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10125800 Supplier Thermo Scientific Acros Supplier No. 307360010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 25597-16-4
Color Colorless
Density 1.1200g/mL
Boiling Point 114.0°C to 115.0°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H7F3O2
Linear Formula CF3CH=CHCO2C2H5
Refractive Index 1.3591 to 1.3611
MDL Number MFCD00009903
Quantity 1 g
Specific Gravity 1.12
Synonym ethyl 4,4,4-trifluorocrotonate, ethyl 2e-4,4,4-trifluorobut-2-enoate, ethyl4,4,4-trifluorocrotonate, e-ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluorocrotonic acid ethyl ester, ethyl e-4,4,4-trifluorobut-2-enoate, ethyl-4,4,4-trifluorocrotonate, ethyl 4,4,4-trifluorobut-2-enoate, 4,4,4-trifluoro-but-2-enoic acid ethyl ester, 2-butenoic acid, 4,4,4-trifluoro-, ethyl ester, 2e
InChI Key ZKRJCMKLCDWROR-ONEGZZNKSA-N
SMILES CCOC(=O)\C=C\C(F)(F)F
IUPAC Name ethyl (2E)-4,4,4-trifluorobut-2-enoate
Molecular Weight (g/mol) 168.12
PubChem CID 5371261
Formula Weight 168.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Ethyl 4,4,4-trifluorocrotonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.