missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-dichloroquinoline-3-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15445478
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15445478 1 g 1g
15494896 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15445478 Supplier Thermo Scientific Alfa Aesar Supplier No. H59231.03
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21168-41-2 | Molecular Formula: C12H9Cl2NO2 | Molecular Weight: 270.11 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Notes
Store at room temperature. Keep away from oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21168-41-2
Molecular Formula C12H9Cl2NO2
Molecular Weight (g/mol) 270.11
MDL Number MFCD00173348
InChI Key BSSNTDZRVNQGDF-UHFFFAOYSA-N
Synonym ethyl 4,6-dichloro-3-quinolinecarboxylate, 4,6-dichloroquinoline-3-carboxylic acid ethyl ester, ethyl-4,6-dichloroquinoline-3-carboxylate, 3-quinolinecarboxylic acid, 4,6-dichloro-, ethyl ester, ethyl-4, 6-dichloroquinoline-3-carboxylate, 4,6-dichloro-3-quinolinecarboxylic acid ethyl ester, 3-quinolinecarboxylicacid, 4,6-dichloro-, ethyl ester, 4,6-dichloroquinoline-3-carboxylic acid, ethyl ester
PubChem CID 1479087
IUPAC Name ethyl 4,6-dichloroquinoline-3-carboxylate
SMILES CCOC(=O)C1=CN=C2C=CC(Cl)=CC2=C1Cl

Specifications

Quantity 1 g
Formula Weight 270.11
Percent Purity 97%
Chemical Name or Material Ethyl 4,6-dichloroquinoline-3-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.