missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 98%

Product Code. 11463607
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463607 250 g 250g
11443607 10 g 10g
11453607 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463607 Supplier Thermo Scientific Alfa Aesar Supplier No. A17365.30
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2386-26-7 | C11H15NO3 | 209.25 g/mol

It is utilized as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is utilized as pharmaceutical intermediate.

Solubility
Slightly soluble in water. 

Notes
Refrigerated (2-4°C). Keep away from strong oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-26-7
Molecular Formula C11H15NO3
Molecular Weight (g/mol) 209.25
MDL Number MFCD00022373
InChI Key ALRDOFWBPAZOCW-UHFFFAOYSA-N
Synonym 3-acetyl-2,4-dimethyl-5-carbethoxypyrrole, 2,4-dimethyl-3-acetyl-5-carbethoxypyrrole, 1h-pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate, 4-acetyl-3,5-dimethyl-pyrrole-2-carboxylic acid ethyl ester, 3-acetyl-2,4-dimethyl-5-ethoxycarbonyl pyrrole, pyrrole-2-carboxylic acid, 4-acetyl-3,5-dimethyl-, ethyl ester, 4-acetyl-3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester, maybridge1_002117, acmc-1cg5g
PubChem CID 16947
IUPAC Name ethyl 4-acetyl-3,5-dimethyl-1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=C(C)C(C(C)=O)=C(C)N1

Specifications

Melting Point 140°C to 143°C
Quantity 250 g
Solubility Information Slightly soluble in water.
Formula Weight 209.25
Percent Purity 98%
Chemical Name or Material Ethyl 4-acetyl-3,5-dimethylpyrrole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.