missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-Amino-3-hydroxybenzoate, TRC

Product Code. 30381583
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381583 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30381583 Supplier Toronto Research Chemicals Supplier No. TRCE89832010G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 4-Amino-3-hydroxybenzoate
CAS 87081-52-5
Molecular Formula C9H11NO3
Quantity 10 g
Synonym 4-Amino-3-hydroxy-benzoic Acid Ethyl Ester, 4-Amino-3-hydroxybenzoic Acid Ethyl Ester
InChI Formula InChI=1S/C9H11NO3/c1-2-13-9(12)6-3-4-7(10)8(11)5-6/h3-5,11H,2,10H2,1H3
SMILES CCOC(=O)c1ccc(N)c(O)c1
Molecular Weight (g/mol) 181.19
Formula Weight 181.0739
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.