missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-chloro-6-hydroxy-5-nitronicotinate, 97%, Thermo Scientific™

Product Code. 15478355
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15478355

Brand: Thermo Scientific Alfa Aesar H33192.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1210835-74-7
Molecular Formula C8H7ClN2O5
Molecular Weight (g/mol) 246.603
MDL Number MFCD22989339
InChI Key FETZVQJUBMRFJE-UHFFFAOYSA-N
Synonym ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydropyridine-3-carboxylate, ethyl 4-chloro-6-hydroxy-5-nitronicotinate, ethyl 4-chloro-6-hydroxy-5-nitropyridine-3-carboxylate, ethylchloronitrooxodihydropyridinecarboxylate, 4-chloro-6-hydroxy-5-nitronicotinic acid ethyl ester, ethyl 4-chloro-5-nitro-6-oxo-1,6-dihydro-3-pyridinecarboxylate, 4-chloro-5-nitro-6-oxo-1,6-dihydro-pyridine-3-carboxylic acid ethyl ester
PubChem CID 59225818
IUPAC Name ethyl 4-chloro-5-nitro-6-oxo-1H-pyridine-3-carboxylate
SMILES CCOC(=O)C1=CNC(=O)C(=C1Cl)[N+](=O)[O-]

Specifications

Melting Point 176°C to 178°C
Color Pink to White
Quantity 250 mg
Formula Weight 246.6
Percent Purity 97%
Physical Form Solid
Chemical Name or Material Ethyl 4-chloro-6-hydroxy-5-nitronicotinate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.